Pipe Stem Triangles
Pipe Stem Triangle, side length 4 cm (1.5")
Catalog # 470189-154
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: CPT015
Jump to:
Product Details & Documents
Triangles made from galvanized iron wire, with porcelain pipe stemming to reduce wire sagging and oxidation.
- Heavy gauge wire
- Galvanized steel
Provides a patform to hold items over a Bunsen burner.
Especificaciones
- Description:Pipe Stem Triangle, side length 4 cm (1.5")