Orders LinkContactUsLinkComponent
LaMotte® Phosphate Test Kit
Water Test Kit, High Range Phosphate, 50 tests
  470021-186
 :  4408-01
LaMotte® Phosphate Test Kit
  470021-186
LaMotte® Phosphate Test Kit
LaMotte® Phosphate Test Kit
Water Test Kit, High Range Phosphate, 50 tests
$4,600.39
Each (1UN)
Precalibrated test tubes and labware give fast, accurate results on-site, in the classroom, or in the lab.
  • Range 1.0 - 10.0 ppm (PO4)
  • Range 10 - 100 ppm (PO4)
Reagents, labware, and step-by-step instructions are furnished in a compact, waterproof carrying case. Kit contains materials for 50 tests.
  • Description:
    Water Test Kit, High Range Phosphate
  • No. of Parts:
    50 tests
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.