Peridinium Slide
Peridinium, Whole mount, Fast Green and Mallory's
470181-020
: Avantor
Plated Cell Walls, Two Flagella
- Identifying features clearly distinguishable
- Rigorous quality control standards
Freshwater. High concentrations cause odor issues in drinking water.
- Description:Peridinium
- Stain:Fast Green and Mallory's
- Orientation:Whole mount