Penicillium Slide
Penicillium Slide, Section, Quadruple
Catalog # 470181-068
Supplier: Avantor
Jump to:
Product Details & Documents
Mold structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
Orange skin showing mycelia, conidiophores, and conidia.
Specifications
- Description:Penicillium Slide
- Stain:Quadruple
- Orientation:Section