Peak Loupe
Peak Loupe
Catalog # 470021-810
Supplier: CHADWICK OPTICAL INC SE
Supplier #: 1962
Jump to:
Product Details & Documents
Large Lenses for Wide, Distortion-Free Viewing Area
The clear plastic base of this convenient loupe can be placed over a live specimen to contain it for closer examination. Size: 1 1/4"L x 1 3/8" widest diameter. 15X magnification.
Specifications
- Magnification:15×
- Description:Peak Loupe