Orders LinkContactUsLinkComponent
Banana Plug Patch Cords
Banana Plug Patch Cords
Banana Plug Patch Cords
Banana Plug Patch Cords
Double ended banana plug jumper wires.
  • Available in red or black
  • 15 cm (6") or 61 cm (24") long
Stackable 4 mm banana plugs on both ends of these flexible, plastic coated wires. The wires have a rating of 15 A, AC.
Product Family Options
  • Description
  • Patch cord, black, 15 cm, banana plugs both ends
    Each (1UN)
    Each (1UN) - $52.56
    $52.56
    Paquete de 6 (1UN)
    Paquete de 6 (1UN) - $57.29
    $57.29
    Each (1UN)
       $52.56
    Paquete de 6 (1UN)
       $57.29
     : 1350302190001
  • Patch cord, red, 15 cm, banana plugs both ends
    Each (1UN)
    Each (1UN) - $52.56
    $52.56
    Paquete de 6 (1UN)
    Paquete de 6 (1UN) - $57.29
    $57.29
    Each (1UN)
       $52.56
    Paquete de 6 (1UN)
       $57.29
     : 1350302190002
  • Patch cord, black, 61 cm, banana plugs both ends
     : 1350302150001
  • Patch cord, red, 61 cm, banana plugs both ends
    Each (1UN)
    Each (1UN) - $61.21
    $61.21
    Paquete de 6 (1UN)
    Paquete de 6 (1UN) - $66.72
    $66.72
    Each (1UN)
       $61.21
    Paquete de 6 (1UN)
       $66.72
     : 1350302150002
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.