Orders LinkContactUsLinkComponent
Partial Immersion Alcohol Thermometers
Partial Immersion Alcohol Thermometers
Partial Immersion Alcohol Thermometers
Certificates
Partial Immersion Alcohol Thermometers
Partial Immersion Alcohol Thermometers
3 Options Available Below
Product Details & Documents
Ideal for most school laboratory applications
  • Feature general-purpose temperature ranges
  • Includes ring top and no-roll sleeve
Product Family Options
Información del producto
  • Range
    Length
    Divisions
    Background Color
    Immersion
    Availability
    Price
  • 0...+230 °F
    30 cm (12")
    2 °F
    White
    Partial (76 mm)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $86.31
    $86.31
    Log in Para disponibilidad
    Caja de 10 (1UN)
    Caja de 10 (1UN) - $94.07
    $94.07
    Each (1UN)
       $86.31
    Caja de 10 (1UN)
       $94.07
    Supplier Number: S230FW-3S
  • −20…+110 °C (0...230 °F)
    30 cm (12")
    1 °C
    Yellow
    Partial (76 mm)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $57.03
    $57.03
    Each (1UN)
       $57.03
    Supplier Number: 1350802410004
  • 470126-656
    Partial immersion alcohol thermometer
    Partial immersion alcohol thermometer
    −20…+110 °C
    30 cm (12")
    1 °C
    White
    Partial (76 mm)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $56.51
    $56.51
    Each (1UN)
       $56.51
    Supplier Number: 1350802310003