Parotid Gland, Human Slide
Parotid Gland, Human, Section, Hematoxylin and Eosin
Catalog # 470183-040
Supplier: Avantor
Jump to:
Product Details & Documents
Major Salivary Gland
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Shows serous salivary acini, ducts, interlobular connective tissue, and fat cells.
Especificaciones
- Description:Parotid Gland, Human
- Stain:Hematoxylin and Eosin
- Orientation:Section