Orders LinkContactUsLinkComponent
<i>Paramecium</i>, Four Types Slide
Paramecium, Four Types Slide
Paramecium, Four Types, Explano-Mount™, Carmine and Fast Green
Catalog # 470181-470
Supplier:   Avantor
<i>Paramecium</i>, Four Types Slide
Catalog # 470181-470
Supplier:  Avantor
<i>Paramecium</i>, Four Types Slide
Paramecium, Four Types Slide
Paramecium, Four Types, Explano-Mount™, Carmine and Fast Green
Price
$135.18
Log in for availability
Each (1UN)
Product Details & Documents
Compare and Contrast Various Paramecia
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Holotrichs. Mixed forms for comparison: P. aurelia, P. bursaria, P. caudatum, and P. multimicronucleatum.
Specifications
  • Description:
    Paramecium, Four Types
  • Stain:
    Carmine and Fast Green
  • Orientation:
    Explano-Mount™