Pandorina Slide
Pandorina, Whole mount, Fast Green and Mallory's
Catalog # 470180-980
Supplier: Avantor
Jump to:
Product Details & Documents
Genus of Green Algae
- Identifying features clearly distinguishable
- Rigorous quality control standards
Spherical, 16-celled colony.
Especificaciones
- Description:Pandorina
- Stain:Fast Green and Mallory's
- Orientation:Whole mount