Pacinian Corpuscle Slide
Pacinian Corpuscle Slide, Whole mount, Carmine
Catalog # 470182-700
Supplier: Avantor
Jump to:
Product Details & Documents
Sensory nerve that senses pressure and vibrations
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Vater’s (Pacinian) corpuscles from mesentery. It is an example of an encapsulated nerve ending.
Especificaciones
- Description:Pacinian Corpuscle Slide
- Stain:Carmine
- Orientation:Whole mount