Orders LinkContactUsLinkComponent
Ward's® Owls and Owl Pellets Reference Poster
Ward's® Owls and Owl Pellets Reference Poster
Owls and Owl Pellets Reference Poster, 38", 27"
Catalog # 470023-238
Supplier:   Avantor
Ward's® Owls and Owl Pellets Reference Poster
Catalog # 470023-238
Supplier:  Avantor
Ward's® Owls and Owl Pellets Reference Poster
Ward's® Owls and Owl Pellets Reference Poster
Owls and Owl Pellets Reference Poster, 38", 27"
Price
$1,703.40
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Colorful Visual Companion to Lessons on Owls and Owl Pellet Dissections
Point out habits and characteristics of owls, and the contents of owl pellets, all featured on this unique, comprehensive poster. It’s an ideal supplement to lectures and a handy reference for students as they conduct owl pellet dissection investigations; the detailed prey identification key helps students recognize the materials they’ll find in their owl pellets. Size: 27" x 38".
Especificaciones
  • Width:
    38"
  • Description:
    Owls and Owl Pellets Reference Poster
  • Height:
    27"