Oversized Demonstration Barrel
Oversized Demonstration Barrel, 5", 1"
Catalog # 470023-278
Supplier: PRECISION FORENSICS TESTING
Supplier #: BL-65
Jump to:
Product Details & Documents
Up-Close view Without a Microscope
Demonstrate barrel characteristics without the need for magnification. This enlarged replica of a firearm barrel
demonstrates rifling, so the entire class can clearly see. The oversized aluminum barrel has been cut in half to
expose large lands and grooves to illustrate key ballistics concepts. Size: 1" dia. x 5"L.
Specifications
- Description:Oversized Demonstration Barrel
- Height:1"
- Width:5"