Osteogenesis, Intramembranous Ossification Slide
Osteogenesis, Intramembranous Ossification, Section, Mallory Triple
Catalog # 470182-650
Supplier: Avantor
Jump to:
Product Details & Documents
Bone Development
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Demonstrates ossification of the skull and the flat bones of face.
Especificaciones
- Description:Osteogenesis, Intramembranous Ossification
- Stain:Mallory Triple
- Orientation:Section