Osteogenesis, Endochondral Ossification Slide
Osteogenesis, Endochondral Ossification Slide, Longitudinal section, Mallory Triple
470176-956
: Avantor
Fetal skeletal development
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal long bone. Extensive deposition of bony spicules with both per- and endochondral osteoblasts and osteoclasts.
- Description:Osteogenesis, Endochondral Ossification Slide
- Stain:Mallory Triple
- Orientation:Longitudinal section