Orders LinkContactUsLinkComponent
Lab-Aids® Osmosis And Diffusion Kit
Lab-Aids® Osmosis And Diffusion Kit
Osmosis And Diffusion Kit, 30
Catalog # 470110-730
Número de proveedor #:  22
Lab-Aids® Osmosis And Diffusion Kit
Catalog # 470110-730
Lab-Aids® Osmosis And Diffusion Kit
Lab-Aids® Osmosis And Diffusion Kit
Osmosis And Diffusion Kit, 30
Price
$5,243.73
Log in Para disponibilidad
Kit de 1
Product Details & Documents
Observe the movement of water and solutes.
  • Observe diffusion through a membrane
  • Time required: one 50 minutes class period
  • Color changes help students visualize osmosis
Can studying the basic life process function of osmosis and diffusion stimulate your students' curiosity? It can with this hands-on lab experience! Students observe first hand the characteristics of a deferentially permeable membrane. Some substances will pass through the membrane and some won't. Some will actually pass through the membrane in both directions simultaneously. Simple color changes help students visualize this biological-physiological phenomenon.
Delivery information: See datasheet for contents information.
Documents
Especificaciones
  • Description:
    Osmosis And Diffusion Kit
  • No. of Lab Groups:
    30