- Descripción:Organic/Inorganic Teacher Set, 232 Pieces
- Referencia:470149-218
- Nº de proveedor:SET00609
Demonstrate Atomic Bonding Using Eight Different Elements
Use this colorful visual aid to lead a demonstration of basic principles of organic chemistry, atomic bonding, and molecular structure in your high school or college-level course. Includes color-coded balls for carbon, hydrogen, nitrogen, oxygen, sulfur, phosphorus, halogen and metal elements with holes for inserting links. Also includes 12 orbitals, and both flexible and hard bonds. Spheres range in diameter from 15mm to 22mm.
Contents:
Qty Element Color Holes Diameter
16 Carbon Black 4 22 mm
6 Carbon Black 3 22 mm
14 Hydrogen White 1 15 mm
2 Hydrogen White 2 20 mm
6 Nitrogen Blue 4 22 mm
4 Nitrogen Blue 3 22 mm
14 Oxygen Red 2 20 mm
6 Oxygen Red 4 22 mm
6 Oxygen Red 1 15 mm
8 Sulfur Yellow 2 22 mm
4 Sulfur Yellow 4 22 mm
1 Sulfur Yellow 6 22 mm
4 Phosphorus Purple 4 22 mm
1 Phosphorus Purple 5 22 mm
2 Phosphorus Purple 3 22 mm
10 Halogen Green 1 20 mm
4 Metal Grey 1 15 mm
3 Metal Grey 2 22 mm
6 Metal Grey 4 22 mm
1 Metal Grey 6 22 mm
6 Orbitals-2D Pink
6 Orbitals-2D Grey
50 Bonds Grey
38 Flexible Long Bonds Grey
12 Bonds Purple