Olfactory Epithelium Slide
Olfactory Epithelium Slide, Section, Hematoxylin and Eosin
Catalog # 470182-744
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates specialized nasal tissue
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Shows pseudostratified columnar epithelium with no goblet cells and no distinct basal lamina.
Especificaciones
- Description:Olfactory Epithelium Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section