Orders LinkContactUsLinkComponent
Ohaus® Spring Scales
Ohaus® Spring Scales
Supplier:   OHAUS CORP.
Ohaus® Spring Scales
Supplier:  OHAUS CORP.
Certificates
Ohaus® Spring Scales
Ohaus® Spring Scales
8 Options Available Below
Product Details & Documents
These spring scales are designed to stand up to heavy student use.
  • Scales withstand sudden load release
  • Individually calibrated
  • Color-coded
  • High-contrast markings
Each scale has been individually calibrated for accuracy and can be fully zeroed. They withstand sudden load releases without breaking.
Scales are color coded, and high-contrast markings make readings easy.
Product Family Options
Dual Scale Metric/Newton
Información del producto
  • Weighing Capacity
    Availability
    Price
  • 470005-236
    8001-MN - 10 g (0.1 N) graduation intervals
    8001-MN - 10 g (0.1 N) graduation intervals
    250 g (2.5 N)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $938.10
    $938.10
    Each (1UN)
       $938.10
    Supplier Number: 80000014
  • 470005-238
    8002-MN - 20 g (0.2 N) graduation intervals
    8002-MN - 20 g (0.2 N) graduation intervals
    500 g (5 N)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $938.10
    $938.10
    Each (1UN)
       $938.10
    Supplier Number: 80000015
  • 470005-240
    8003-MN - 25 g (0.25 N) graduation intervals
    8003-MN - 25 g (0.25 N) graduation intervals
    1000 g (10 N)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $938.10
    $938.10
    Each (1UN)
       $938.10
    Supplier Number: 80000016
  • 470005-234
    8004-MN - 50 g (0.5 N) graduation intervals
    8004-MN - 50 g (0.5 N) graduation intervals
    2000 g (20 N)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $938.10
    $938.10
    Each (1UN)
       $938.10
    Supplier Number: 80000017
  • 470203-702
    8008-MN - 100 g (1 N) graduation intervals
    8008-MN - 100 g (1 N) graduation intervals
    5000 g (50 N)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $938.10
    $938.10
    Each (1UN)
       $938.10
    Supplier Number: 80000018
Dual Scale Metric/English
Información del producto
  • Weighing Capacity
    Availability
    Price
  • 470203-700
    8001-MA - 10 g (0.25 oz.) graduation intervals
    8001-MA - 10 g (0.25 oz.) graduation intervals
    250 g (9 oz.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,026.04
    $1,026.04
    Each (1UN)
       $1,026.04
    Supplier Number: 80000039
  • 470006-148
    8002-MA - 20 g (0.50 oz.) graduation intervals
    8002-MA - 20 g (0.50 oz.) graduation intervals
    500 g (18 oz.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,026.04
    $1,026.04
    Each (1UN)
       $1,026.04
    Supplier Number: 80000041
Dual Scale Pound/Newton
Información del producto
  • Weighing Capacity
    Availability
    Price
  • 470004-632
    8002-PN - 0.04 lbs. (0.2 N) graduation intervals
    8002-PN - 0.04 lbs. (0.2 N) graduation intervals
    1.12 lbs. (5 N)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,114.00
    $1,114.00
    Each (1UN)
       $1,114.00
    Supplier Number: 80000042