Plastic Flower Pots
Flower Pot, 12.5 cm (5 in)
Catalog # 470211-884
Supplier: Avantor
Jump to:
Product Details & Documents
High-impact plastic makes these plant pots ideal for classroom use.
- The economical and attractive pots are easy to clean and are also reusable
Packaging: Packages of 10.
Especificaciones
- Description:Flower Pot, 12.5 cm (5 in)