North American Rock Collection
470151-158
- Descripción:North American Rock Collection
- No. of Samples:100
- Referencia:470151-158
- Nº de proveedor:7070P-121
This collection with rock types and associations with the North American continent contains 100 specimens with each group represented: igneous, metamorphic, and sedimentary. Specimens are approximately 1"" x 1"", number-coded in compartmented container.
Key also contains specimen locality information.