Non-Scratching Duster for Lab Equipment
Brush Duster, Non-Scratching, Synthetic fibres, Yellow, 4 cm, Plastic, White, 12 cm
470006-808
: B265C
Non-scratching duster for microscopes, laboratory equipment, printed circuits and other delicate equipment and instruments.
- Bristle Color:Yellow
- Bristle Material:Synthetic fibres
- Handle Material:Plastic
- Description:Brush Duster, Non-Scratching
- Handle Length:12 cm
- Handle Color:White
- Brush Head Length:4 cm