Nickel/Stainless Steel Spatulas, Double Bladed
Proveedor: VWR International
Certificados
Acerca de este artículo
Spatulas with 50×7 mm thin flat rounded blades on either end.
Wire Ø: 3 mm
Length: 8"
Frequently Bought Together
Spatulas with 50×7 mm thin flat rounded blades on either end.
Wire Ø: 3 mm
Length: 8"