Orders LinkContactUsLinkComponent
Curriculum Mastery Posters
Curriculum Mastery Posters
Curriculum Mastery Posters
Certificates
Curriculum Mastery Posters
Curriculum Mastery Posters
5 Options Available Below
Product Details & Documents
A Graphic Look At Key Science Concepts
  • Detailed illustrations
  • Representative photographs
  • Colorful depictions
Large, reference posters facilitate comprehension of key, standards-based concepts. Each poster is laminated for years of use.
Cells
Vivid images compare and contrast the similarities and differences between plant and animal cells, including detailed representation of each organelle and examples of various specialized cells.
Osmosis and Diffusion
Examine the structure of cell membranes, the processes of diffusion and osmosis and the differences between them.
Modern Genetics
Thorough, visual overview of the structure and function of chromosomes, genes and DNA also provides a step-by-step procedure for DNA fingerprinting.
Photosynthesis and Respiration
Take a comprehensive look at the steps of photosynthesis and the two stages of respiration.
Microlife
Illustrations and photographs show a typical bacterial cell, various fungi types, as well as amoeba, paramecium and euglena.
Product Family Options
Información del producto
  • Finish
    Length
    Width
    Availability
    Price
  • Cells
    Laminated
    89 cm
    58.5 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $563.25
    $563.25
    Log in Para disponibilidad
    Paquete de 20 (1UN)
    Paquete de 20 (1UN) - $613.94
    $613.94
    Each (1UN)
       $563.25
    Paquete de 20 (1UN)
       $613.94
    Supplier Number: 346101
  • 470144-526
    Osmosis And Diffusion
    Osmosis And Diffusion
    Laminated
    89 cm
    58.5 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $563.25
    $563.25
    Log in Para disponibilidad
    Paquete de 20 (1UN)
    Paquete de 20 (1UN) - $613.94
    $613.94
    Each (1UN)
       $563.25
    Paquete de 20 (1UN)
       $613.94
    Supplier Number: 346102
  • 470144-612
    Modern Genetics
    Modern Genetics
    Laminated
    89 cm
    58.5 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $563.25
    $563.25
    Log in Para disponibilidad
    Paquete de 20 (1UN)
    Paquete de 20 (1UN) - $613.94
    $613.94
    Each (1UN)
       $563.25
    Paquete de 20 (1UN)
       $613.94
    Supplier Number: 346103
  • 470144-670
    Mitosis And Meiosis
    Mitosis And Meiosis
    Laminated
    89 cm
    58.5 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $563.25
    $563.25
    Log in Para disponibilidad
    Paquete de 20 (1UN)
    Paquete de 20 (1UN) - $613.94
    $613.94
    Each (1UN)
       $563.25
    Paquete de 20 (1UN)
       $613.94
    Supplier Number: 346104
  • 470144-522
    Photosynthesis And Respiration
    Photosynthesis And Respiration
    Laminated
    89 cm
    58.5 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $563.25
    $563.25
    Log in Para disponibilidad
    Paquete de 20 (1UN)
    Paquete de 20 (1UN) - $613.94
    $613.94
    Each (1UN)
       $563.25
    Paquete de 20 (1UN)
       $613.94
    Supplier Number: 346105