Specifications
- Descripción:Owls and owl pellets flip chart
- Altura:12"
- Referencia:470144-502
- Anchura:18"
- Nº de proveedor:346015
Specifications
About this item
Interactive, hands-on, and digital resources about owls, owl pellet dissection, and prey identification.
- Excellent alternative or supplementary activity to actual owl pellets
- Blended learning activities for interactive group learning
- Bundled and cost-effective purchasing opportunities
Owl pellet virtual dissection curriculum module: Used in combination or individually, your students will gain a thorough comprehension of owl pellet dissection through an engaging blend of interactive print resources combined with online lessons and activities - including an owl pellet dissection virtual lab. The perfect substitute for owl pellet dissection lessons or for pre-lab prep or post-lab review when conducting actual hands-on labs.
Module includes - One owls and owl pellets flip chart set, eight owl pellet dissection resource guides, and one year online access for teacher and all of their students to an online lesson featuring an owl pellet dissection virtual lab, narrated tutorials, interactive exercises, quizzes and more.
Owls and owl pellets flip chart set with online multimedia lesson: Engaging, easy-to-use graphic resource features ten double-sided, laminated 18×12" charts. one side features an illustrated overview of the topic while the reverse provides write-on/wipe-off exercises for review and reinforcement. spiral-bound on a durable, free-standing easel, the set also includes an activity guide with copymasters of review questions and exercises. also available with one-year access to a complementary online lesson featuring owl pellet dissection virtual lab, narrated tutorials, interactive exercises, quizzes and more!
Set includes ten doubled-sided charts - owls – birds of prey; the barn owl; owl food web; owl pellets; dissecting an owl pellet; rodent skeleton; vole skeleton; mole skeleton; shrew skeleton; and bird skeleton.
Owls and owl pellet dissection resource guide with online multimedia lesson: full-color, comprehensive overview of owl pellet dissection and prey identification! features self-directed reading, easy-to-follow illustrated explanations, guiding questions, inquiry-based activities, a lab investigation, key vocabulary review, assessment review questions and a post-test. also available with one-year access to a complementary online lesson featuring owl pellet dissection virtual lab, narrated tutorials, interactive exercises, quizzes and more!
Covers: owls – birds of prey; the barn owl; owl food web; all about pellets; rodent identification; vole identification; mole identification; shrew identification; bird identification; and dissecting an owl pellet lab investigation. 40 pages.