Nervous Tissue, Peripheral Nerve Slide
Nervous Tissue, Peripheral Nerve Slide, Cross section, Hematoxylin and Eosin
Catalog # 470182-694
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates mammalian nervous tissue
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal medullated nerve.
Especificaciones
- Description:Nervous Tissue, Peripheral Nerve Slide
- Stain:Hematoxylin and Eosin
- Orientation:Cross section