Muscle Types, Composite, Human Slide
Muscle Types, Composite, Human Slide, Section, Hematoxylin and Eosin
Catalog # 470177-008
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates the differences between muscle types
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Smooth, cardiac, and skeletal muscle.
Especificaciones
- Description:Muscle Types, Composite, Human Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section