Multiple Roll Tape Dispenser, 1" Core
Multiple roll tape dispenser steel 1" Core
Catalog # 470018-038
Supplier: NEVS INK INC SE
Número de proveedor #: VWR-EDU-PT
Jump to:
Product Details & Documents
Designed for tape with 1" cores, this dispenser accommodates 1/2", 3/4", 1", 1 1/2", or 2" wide tapes for a total dispensing width of 9 1/2". Dispenser also features bottom grips to stay securely in place during use.
Especificaciones
- Description:Multiple roll tape dispenser steel 1" Core