Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Especificaciones
- Descripción:Mr. Mole Kit
- No. of Lab Groups:6
- Referencia:470024-326
- Nº de proveedor:6-100
Especificaciones
Acerca de este artículo
Dig into Avogadro’s Number
- Allows students to discover the most important unit of substance in chemistry
In this kit students will determine the molar mass of various substances and then express it in grams. Finally, using Avogadro’s number, they will learn to convert moles to molecules. The kit even includes a pattern for a mole puppet and mole jokes, to make the learning experience fun and memorable.