Orders LinkContactUsLinkComponent
<i>Mosasaurus</i> sp. (Cretaceous)
Mosasaurus sp. (Cretaceous)
Mosasaurus sp. (Cretaceous) Fossil Reproduction, 17", 32", 2¼"
Catalog # 470237-662
Supplier:   Avantor
<i>Mosasaurus</i> sp. (Cretaceous)
Catalog # 470237-662
Supplier:  Avantor
<i>Mosasaurus</i> sp. (Cretaceous)
Mosasaurus sp. (Cretaceous)
Mosasaurus sp. (Cretaceous) Fossil Reproduction, 17", 32", 2¼"
Price
$6,828.55
Log in for availability
Each (1UN)
Product Details & Documents
Skull replica.
  • Chordata
This large, well-prepared skull replica is from an ancient marine reptile that swam the seas of the Kansas Seaway some 92 million years ago. Its total body length was nearly 15 to 20 feet, making it one of the largest marine predators of its time. Kansas.
Specifications
  • Description:
    Mosasaurus sp. (Cretaceous) Fossil Reproduction
  • Height:
    2¼"
  • Length:
    32"
  • Width:
    17"