Economy Mortar & Pestle Sets
Supplier: UNITED SCIENTIFIC SUPPLIES
Certificates
About this item
Quickly grind materials to a powder.
- Glazed on the outside
- Pouring spout
Affordable student quality sets with pour-out lips. Designed for general laboratory use, the mortars are glazed on the outside and unglazed on the inside.
Frequently Bought Together