Molded Plastic Case
Molded Plastic Case
Catalog # 470147-518
Supplier: ONTARIO PLASTICS INC
Número de proveedor #: 470147-518
Jump to:
Product Details & Documents
Protect sharp instruments, and yourself, with this sturdy case of injection-molded polypropylene.
- Assemble Your Own Dissection Kits
- Prevents Cuts Or Punctures While Carrying Sharp Instruments
- Store Instruments Neatly While Not In Use
Lifetime hinges, firm latches, and six instrument compartments are built in.
Especificaciones
- Description:Molded Plastic Case