Modeling Stream Erosion and Deposition Kit
Proveedor: LAB-AIDS INC (KITS) SE
Certificados
Acerca de este artículo
Ideal Size For Student Demos
- Demonstrated formation of landforms
- Teachers guide included
Investigate the dynamics of stream erosion and deposition with this inventive miniature model. Using specially engineered materials, students create, observe, describe, and interpret the erosional and depositional patterns formed when rain water flows over an area. Some common landform features include canyons, braided streams, and deltas. In the process, students learn that water falls to the surface where it collects in rivers, lakes, and oceans. Includes a teacher’s guide and student worksheets.
Frequently Bought Together