Mixed Protist Slide
Mixed Protist Slide, Explano-Mount™, Fast Green and Mallory's
Catalog # 470177-490
Supplier: Avantor
Jump to:
Product Details & Documents
Depicts protist diversity
- Identifying features clearly distinguishable
- Rigorous quality control standards
Many examples from three major phyla: ciliates, flagellates, and amoebae.
Especificaciones
- Description:Mixed Protist Slide
- Stain:Fast Green and Mallory's
- Orientation:Explano-Mount™