Orders LinkContactUsLinkComponent
Mixed Pollen, 20 Types Slide
Mixed Pollen, 20 Types Slide
Mixed Pollen, 20 Types, Whole mount, Composite: Multiple stains
Catalog # 470181-214
Supplier:   Avantor
Mixed Pollen, 20 Types Slide
Catalog # 470181-214
Supplier:  Avantor
Mixed Pollen, 20 Types Slide
Mixed Pollen, 20 Types Slide
Mixed Pollen, 20 Types, Whole mount, Composite: Multiple stains
Price
$135.75
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Identify Various Pollen by its Form
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Monocot, dicot, and gymnosperm. A key card is supplied.
Especificaciones
  • Description:
    Mixed Pollen, 20 Types
  • Stain:
    Composite: Multiple stains
  • Orientation:
    Whole mount