Mixed Green Algae Slide
Mixed Green Algae Slide, Whole mount, Fast Green and Mallory's
470177-430
: Avantor
Illustrates a variety of green algae forms
- Identifying features clearly distinguishable
- Rigorous quality control standards
Includes six types. Examples of filamentous, branched, colonial, and single celled genera shown.
- Description:Mixed Green Algae Slide
- Stain:Fast Green and Mallory's
- Orientation:Whole mount