Mixed Dinoflagellates Slide
Mixed Dinoflagellates Slide, Explano-Mount™, Fast Green and Mallory's
470177-020
: Avantor
Illustrates dinoflagellate diversity
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mixed freshwater and marine forms.
- Description:Mixed Dinoflagellates Slide
- Stain:Fast Green and Mallory's
- Orientation:Explano-Mount™