Mixed Ciliates Slide
Mixed Ciliates, Explano-Mount™, Fast Green and Mallory's
Catalog # 470181-462
Supplier: Avantor
Jump to:
Product Details & Documents
Protozoans Usually Found in Water and Soil
- Identifying features clearly distinguishable
- Rigorous quality control standards
Variety of common species are shown.
Especificaciones
- Description:Mixed Ciliates
- Stain:Fast Green and Mallory's
- Orientation:Explano-Mount™