ContactUsLinkComponent
TeacherGeek Mini Wind Turbine
TeacherGeek Mini Wind Turbine
Catálogo # 470132-396
Proveedor:  TEACHERGEEK INC
undefined
TeacherGeek Mini Wind Turbine
Catálogo # 470132-396
Proveedor:  TEACHERGEEK INC
Número de proveedor:  1823-13
Especificaciones
  • Descripción:
    Mini wind turbine, pack of 10
  • Referencia:
    470132-396
  • Nº de proveedor:
    92101
Especificaciones
Acerca de este artículo
Investigate the Impact of Blade Design on Power Output
  • Easy to connect parts
  • Reusable pieces
  • Hands on way to investigate the design process
  • A true STEM experience
  • Build stators and coils
The TeacherGeek Mini Wind Turbine Activity provides you the opportunity to build a wind powered generator. You will start by building the stand from a hole plate, dowels, perpendicular blocks and hardware. The TeacherGeek uniquely designed hub slides directly on to the motor shaft and securely holds 2,3 or 6 blades. These blades are constructed from found materials mounted with tape on to provided wooden skewers. The DC motor is spun by wind power creating a mini generator. Your innovation comes in to play as you redesign and reconfigure the blades to change power output. A very complete step by step pictorial Mini Wind Lift Guide (available as a free download) will lead you through construction, give you blade design tips and ideas for experimentation.
Información de pedido: Reamer, Easy Cutter, Phillips Screwdriver, and Slip-Joint Pliers are required but not included.