Orders LinkContactUsLinkComponent
Mini Hacksaw
Mini Hacksaw
Mini Hacksaw
Catalog # 470019-474
Número de proveedor #:  80070
Mini Hacksaw
Catalog # 470019-474
Mini Hacksaw
Mini Hacksaw
Mini Hacksaw
Price
$223.89
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This versatile mini hacksaw can be adjusted to cut through varying thicknesses of multiple surfaces including metal, plastic, and wood.
The five included multi-purpose blades, each 6 1/2" long, can be replaced quickly and easily. Size: 12"L x 5 1/2"H.
Especificaciones
  • Description:
    Mini Hacksaw
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.