Mini Compasses
Mini compass, dia: 16 mm
Catalog # 470003-496
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: C1SG16-PK/12
Jump to:
Product Details & Documents
These small 16 mm diameter compasses are perfect for mapping out magnetic fields lines around a compass.
- Highly accurate
- Features a floating needle
These small compasses are ideal for mapping the magnetic field around a bar magnet.
Especificaciones
- Description:Mini compass, dia: 16 mm