Mini Light Socket with Fahnstock Clips
Mini bulb holder, pack of 6, E10 base
Catalog # 470016-940
Supplier: PRECISION MOULDED POLYMERS LTD
Número de proveedor #: AM6080.00006.00000
Mini Light Socket with Fahnstock Clips
Catalog # 470016-940
Supplier: PRECISION MOULDED POLYMERS LTD
Jump to:
Product Details & Documents
Inexpensive yet convenient for all circuit experiments.
Connect bare wires using fahnestock clips
- Fits E10 screw base mini bulbs
This simple bulb holder is an essential accessory for all circuit experiments.
Ordering information: For a complete bulb selection, see our Incandescent Lamps.
Especificaciones
- Description:Mini bulb holder, pack of 6
- Size:E10 base