Micro-Spatulas
Micro-spatulas, one tapered end, one blunt end, 8"
470020-086
Supplier #: SSFT08
These versatile spatulas have different tools on either end; choose the style best suited to your particular needs.
- Stainless steel
Perfect for use with small sample quantities.
Packaging: Packages of 12.
- Description:Micro-spatulas, one tapered end, one blunt end
- Length:8"