Boreal Science Microscope Maintenance Kit
Product Details & Documents
Keep your microscope in ideal working order with these supplies for maintenance and upkeep.
Designed specifically for our full line of microscopes, this kit helps you perform the routine maintenance on your microscopes that keeps it in perfect working order.
: This toolbox includes one 8 oz. bottle of Boreal lens cleaning solution, one No.2 Phillips head screwdriver, one 1 oz. pipet-aid, one plastic dropper, one camel’s hair brush, one 'C' wrench for microscope tension collars, one package of lens paper, one box of small Kimwipes, one toothbrush, One 1.75 oz. tube of grease, one 3×/6× magnifier, one pair of snipe-nose pliers, one hex-wrench set (7 sizes), one precision screwdriver, one pair of straight sharp-point forceps, and one slotted screwdriver (5 cm blade). The Boreal Microscope Manual with detailed instructions and a troubleshooting guide are also included.
: Items packed in heavy-duty vinyl tool roll.
Especificaciones
- Description:Hex wrench set/7