Microscope Focusing Tips Chart
Catalog # 470023-364
Supplier: BIOCAM COMMUNICATIONS INC SE
Specifications
- Descripción:Microscope Focusing Tips Chart
- Referencia:470023-364
- Nº de proveedor:WC03
Specifications
About this item
Hone Your Microscopy Skills
Nine separate insets show step-by-step techniques, as well as things to avoid when using the microscope to observe a specimen. Each inset contains illustrations, photomicrographs, and descriptions that clearly describe the specific steps in the focusing process. The chart is laminated to prevent tearing. Size: 18" x 24".