Glass coverslips
Glass cover slips, 116 Pieces, 22×22 mm, 0.17 - 0.25 mm
Catalog # 470145-876
Supplier: C & A SCIENTIFIC CO INC
Supplier #: 94-22222
Jump to:
Product Details & Documents
Finest Optical Quality
- Quality slide making supplies
- Traditional coverslip type
These cover glasses are required when using 100X oil immersion objective lenses.
Specifications
- Description:Glass cover slips, 116 Pieces
- L×W:22×22 mm
- Thickness:0.17 - 0.25 mm