Ward's® Microhematocrit Set
WARD’S Microhematocrit Set
Catalog # 470019-794
Supplier: Avantor
Jump to:
Product Details & Documents
The set includes 200 heparinized, mylar wrapped, hematocrit tubes, and six capillary tube trays with sealant. Each tray has two rows of slots numbered 1 through 24 for easy tube storage.
Especificaciones
- Description:WARD’S Microhematocrit Set
- Cat. No.:470019-794