Orders LinkContactUsLinkComponent
Ward's® Microcline (Pink)
Ward's® Microcline (Pink)
Supplier:   Avantor
Ward's® Microcline (Pink)
Supplier:  Avantor
Certificates
Ward's® Microcline (Pink)
Ward's® Microcline (Pink)
4 Options Available Below
Product Details & Documents
Feldspar group; pink cleavages showing perthitic texture, common potassium feldspar, Mohs’ hardness of 6.
  • Collected in Ontario, Canada
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Información del producto
  • Specimen Dimensions
    Availability
    Price
  • 470024-896
    Microcline (Pink) thin section
    Microcline (Pink) thin section
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,045.20
    $1,045.20
    Each (1UN)
       $1,045.20
  • 470025-686
    Microcline (Pink), student specimens
    Microcline (Pink), student specimens
    1×1 - 1×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $315.00
    $315.00
    Paquete de 10
       $315.00
  • 470025-690
    Microcline (Pink), hand specimen, Each
    Microcline (Pink), hand specimen, Each
    2×3" - 3×4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $328.18
    $328.18
    Each (1UN)
       $328.18
  • 470025-688
    Microcline (Pink), bulk pack, 1 kg
    Microcline (Pink), bulk pack, 1 kg
    Size will vary
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $501.73
    $501.73
    Each (1UN)
       $501.73