Microbiology Morphology Slides
Microbiology Morphology Slides, 12, Monera
Catalog # 470176-204
Supplier: Avantor
Jump to:
Product Details & Documents
Investigate the differences between microorganisms
- Identifying features clearly distinguishable
- Rigorous quality control standards
This set is designed to show the basic differential shapes between bacteria and also between bacteria and fungi. An excellent set to demonstrate the morphology of micro-organisms supplied in a durable box.
: Set includes Penicillum (wm); Yeast, budding (wm); Capsules of bacteria sm); Typical coccus (sm); Typical spirillum (sm); Bacteria types (sm); Bacillus in chains (sm); Typical mixed bacteria (sm); Spirochaetes (sm); Treponema pallidium (sm); Typical bacillus (sm); Staphylococcus epidermidis (sm).
Especificaciones
- Type:Monera
- Description:Microbiology Morphology Slides
- No. of Slides:12