Micro, Pure Gum Amber Rubber Tubing
Catalog # 470150-362
Supplier: KENT ELASTOMER PRODUCTS SE
Specifications
- Descripción:Micro, Pure Gum Amber Rubber Tubing
- Referencia:470150-362
- Nº de proveedor:202-12
Specifications
About this item
Small bore with light wall, surgical grade. ID: ¹/₁₆"; OD: ¹/₈"; wall thickness: ¹/₃₂". 12' (3.6 m) length.